Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Loh12cr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170479.1 | NP_001163950.1 | 2 - 3 | 321 | 73 | |
NM_026371.3 | NP_080647.2 | 2 - 3 | 463 | 73 | ||
GenBank mRNA | AK007423.1 | - | 2 - 3 | 320 | 73 | |
AK018019.1 | - | 2 - 3 | 379 | 73 | ||
AK089648.1 | - | 2 - 3 | 327 | 73 | ||
AK151342.1 | - | 2 - 3 | 329 | 73 | ||
AK169615.1 | - | 2 - 3 | 335 | 73 | ||
BC023059.1 | - | 2 - 3 | 422 | 5355560 | 73 | |
DV652098.1 | - | 2 - 3 | 213 | 73 | ||
DV658678.1 | - | 2 - 3 | 323 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |