Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tdrkh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028307.1 | NP_082583.1 | 5 - 6 | 638 | 91 | |
GenBank mRNA | AK012596.1 | - | 5 - 6 | 679 | 91 | |
BC031886.1 | - | 5 - 6 | 532 | 91 | ||
BC049363.1 | - | 5 - 6 | 638 | 6490411 | 91 | |
BC057030.1 | - | 5 - 6 | 654 | 6833743 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |