Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008149.3 | NP_032175.2 | 18 - 19 | 2274 | 76 | |
GenBank mRNA | AK047295.1 | - | 18 - 19 | 2271 | 76 | |
AK137067.1 | - | 20 - 21 | 2555 | 76 | ||
AK173202.1 | - | 18 - 19 | 2299 | 76 | ||
BC019201.1 | - | 18 - 19 | 2279 | 5041120 | 76 | |
BC082776.1 | - | 4 - 5 | 576 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |