Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdzk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001146001.1 | NP_001139473.1 | 5 - 6 | 842 | 77 | |
NM_021517.2 | NP_067492.2 | 5 - 6 | 888 | 77 | ||
GenBank mRNA | AF220100.1 | - | 5 - 6 | 827 | 77 | |
AF474247.1 | - | 4 - 5 | 826 | 76 | ||
AK006269.1 | - | 5 - 6 | 1362 | 77 | ||
AK012070.1 | - | 5 - 6 | 878 | 77 | ||
AK014879.1 | - | 5 - 6 | 1145 | 77 | ||
AK029752.1 | - | 5 - 6 | 1346 | 77 | ||
AK029764.1 | - | 5 - 6 | 1356 | 77 | ||
AK165415.1 | - | 5 - 6 | 823 | 77 | ||
BC013512.1 | - | 4 - 5 | 811 | 4234949 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |