Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0300020022.mm.1 |
NM_001012306.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300020022.mm.1 |
NM_001161742.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300020022.mm.1 |
NM_001161742.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300020022.mm.1 |
NM_001161743.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300020022.mm.1 |
NM_001161743.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300020022.mm.1 |
NM_001161744.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300020022.mm.1 |
NM_001161745.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300020022.mm.1 |
TC0300002559.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300020018.mm.1 |
NM_001012306.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300020018.mm.1 |
NM_001161742.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300020018.mm.1 |
NM_001161742.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300020018.mm.1 |
NM_001161743.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300020018.mm.1 |
NM_001161743.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300020018.mm.1 |
NM_001161744.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300020018.mm.1 |
NM_001161745.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300020018.mm.1 |
TC0300002559.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300020025.mm.1 |
NM_001012306.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300020025.mm.1 |
NM_001161742.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300020025.mm.1 |
NM_001161742.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300020025.mm.1 |
NM_001161743.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300020025.mm.1 |
NM_001161743.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300020025.mm.1 |
NM_001161744.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300020025.mm.1 |
NM_001161745.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300020025.mm.1 |
TC0300002559.mm.1 |
MTA TC |