Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tuft1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001293728.1 | NP_001280657.1 | 5 - 6 | 493 | 79 | |
NM_011656.3 | NP_035786.1 | 6 - 7 | 568 | 79 | ||
GenBank mRNA | AF002860.1 | - | 6 - 7 | 462 | 79 | |
AF047704.1 | - | 6 - 7 | 502 | 79 | ||
AK132488.1 | - | 6 - 7 | 500 | 79 | ||
AK171705.1 | - | 5 - 6 | 458 | 79 | ||
AK206798.1 | - | 132 | 79 | |||
BC019213.1 | - | 6 - 7 | 499 | 5064658 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |