Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pde3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018779.1 | NP_061249.1 | 14 - 15 | 3247 | 83 | |
GenBank mRNA | AF099187.1 | - | 14 - 15 | 3247 | 83 | |
AK052374.1 | - | 14 - 15 | 2523 | 83 | ||
AK164864.1 | - | 14 - 15 | 2030 | 83 | ||
BC138265.1 | - | 14 - 15 | 3205 | 8861284 | 83 | |
BC138266.1 | - | 14 - 15 | 3205 | 8861285 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |