Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aldh7a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127338.1 | NP_001120810.1 | 7 - 8 | 791 | 86 | |
NM_138600.4 | NP_613066.2 | 5 - 6 | 597 | 86 | ||
GenBank mRNA | AK004991.1 | - | 7 - 8 | 725 | 86 | |
AK160117.1 | - | 5 - 6 | 524 | 86 | ||
AK169195.1 | - | 5 - 6 | 540 | 86 | ||
BC012407.1 | - | 5 - 6 | 559 | 4191228 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |