Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phax | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001162989.1 | NP_001156461.1 | 3 - 4 | 861 | 68 | |
NM_019996.4 | NP_064380.3 | 3 - 4 | 971 | 68 | ||
GenBank mRNA | AJ276504.1 | - | 3 - 4 | 802 | 68 | |
AK030749.1 | - | 3 - 4 | 829 | 68 | ||
BC016530.1 | - | 3 - 4 | 818 | 68 | ||
BC049590.1 | - | 3 - 4 | 817 | 6587693 | 68 | |
BC061110.1 | - | 3 - 4 | 831 | 30303763 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons |