Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0100004318.mm.1 |
NM_001037726.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100004318.mm.1 |
NM_001037726.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100004318.mm.1 |
NM_009952.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100004318.mm.1 |
NM_009952.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100004318.mm.1 |
NM_133828.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100004318.mm.1 |
NM_133828.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100004318.mm.1 |
TC0100000535.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100004320.mm.1 |
NM_001037726.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100004320.mm.1 |
NM_001037726.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100004320.mm.1 |
NM_009952.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100004320.mm.1 |
NM_009952.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100004320.mm.1 |
NM_133828.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100004320.mm.1 |
NM_133828.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100004320.mm.1 |
TC0100000535.mm.1 |
MTA TC |