Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400026107.mm.1 |
TC0400003275.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400026107.mm.1 |
NM_001038698.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400026107.mm.1 |
NM_001038698.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400026107.mm.1 |
NM_001163397.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400026107.mm.1 |
NM_001163397.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400026107.mm.1 |
NM_001163399.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400026107.mm.1 |
NM_001163399.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400026107.mm.1 |
NM_010488.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400026107.mm.1 |
NM_010488.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400026106.mm.1 |
TC0400003275.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400026106.mm.1 |
NM_001038698.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400026106.mm.1 |
NM_001038698.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400026106.mm.1 |
NM_001163397.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400026106.mm.1 |
NM_001163397.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400026106.mm.1 |
NM_001163399.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400026106.mm.1 |
NM_001163399.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400026106.mm.1 |
NM_010488.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400026106.mm.1 |
NM_010488.4 |
Refseq |