Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1800006313.mm.1 |
NM_001025245.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800006313.mm.1 |
NM_001025245.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800006313.mm.1 |
NM_010777.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800006313.mm.1 |
NM_010777.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800006313.mm.1 |
TC1800000869.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1800006318.mm.1 |
NM_001025245.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800006318.mm.1 |
NM_001025245.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800006318.mm.1 |
NM_010777.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800006318.mm.1 |
NM_010777.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800006318.mm.1 |
TC1800000869.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1800006314.mm.1 |
NM_001025245.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800006314.mm.1 |
NM_001025245.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800006314.mm.1 |
NM_010777.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800006314.mm.1 |
NM_010777.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800006314.mm.1 |
TC1800000869.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1800006320.mm.1 |
NM_001025245.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800006320.mm.1 |
NM_001025245.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800006320.mm.1 |
NM_010777.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800006320.mm.1 |
NM_010777.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800006320.mm.1 |
TC1800000869.mm.1 |
MTA TC |