Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cxxc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028868.3 | NP_083144.1 | 10 - 11 | 1692 | 68 | |
GenBank mRNA | AK010337.1 | - | 10 - 11 | 1693 | 68 | |
AK083655.1 | - | 10 - 11 | 1697 | 68 | ||
AK146688.1 | - | 10 - 11 | 1776 | 68 | ||
AY099096.1 | - | 10 - 11 | 1664 | 68 | ||
BC012261.1 | - | 6 - 7 | 1235 | 68 | ||
BC030938.1 | - | 10 - 11 | 1524 | 3980339 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |