Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mbd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013594.2 | NP_038622.2 | 1 - 2 | 149 | 86 | |
GenBank mRNA | AF072240.1 | - | 1 - 2 | 152 | 86 | |
AK004624.2 | - | 1 - 2 | 150 | 86 | ||
AK007371.1 | - | 1 - 2 | 833 | 86 | ||
AK156401.1 | - | 1 - 2 | 164 | 86 | ||
AK162858.1 | - | 1 - 2 | 149 | 86 | ||
BC069837.1 | - | 1 - 2 | 165 | 30435760 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |