Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dctn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026302.3 | NP_080578.1 | 7 - 8 | 739 | 65 | |
GenBank mRNA | AK016059.1 | - | 7 - 8 | 793 | 65 | |
AK034292.1 | - | 8 - 9 | 797 | 65 | ||
AK050896.1 | - | 7 - 8 | 743 | 65 | ||
AK141855.1 | - | 8 - 9 | 762 | 65 | ||
AK160813.1 | - | 7 - 8 | 745 | 65 | ||
AK163240.1 | - | 7 - 8 | 761 | 65 | ||
AK168484.1 | - | 7 - 8 | 745 | 65 | ||
BC006677.1 | - | 7 - 8 | 702 | 65 | ||
BC034725.1 | - | 7 - 8 | 739 | 3667031 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |