Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rfx5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017395.2 | NP_059091.2 | 4 - 5 | 365 | 65 | |
GenBank mRNA | AB221679.1 | - | 4 - 5 | 346 | 65 | |
AF209854.1 | - | 4 - 5 | 346 | 65 | ||
AK088686.1 | - | 5 - 6 | 417 | 65 | ||
BB666132.1 | - | 5 - 6 | 391 | 65 | ||
BB854843.1 | - | 5 - 6 | 458 | 66 | ||
BC051965.1 | - | 5 - 6 | 402 | 5702640 | 65 | |
BC057336.1 | - | 5 - 6 | 391 | 65 | ||
BC075717.1 | - | 6 - 7 | 474 | 65 | ||
BC118948.2 | - | 4 - 5 | 346 | 40092396 | 65 | |
BC118949.2 | - | 4 - 5 | 346 | 40092397 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |