Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmod4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016712.3 | NP_057921.1 | 8 - 9 | 975 | 78 | |
GenBank mRNA | AF177174.1 | - | 7 - 8 | 894 | 78 | |
AK144408.1 | - | 8 - 9 | 976 | 78 | ||
AK144535.1 | - | 8 - 9 | 976 | 78 | ||
BC068020.1 | - | 7 - 8 | 911 | 30291625 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |