Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Podn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285956.1 | NP_001272885.1 | 5 - 6 | 895 | 90 | |
NM_001285958.1 | NP_001272887.1 | 5 - 6 | 791 | 90 | ||
NM_172874.3 | NP_766462.2 | 5 - 6 | 817 | 90 | ||
GenBank mRNA | AK034987.1 | - | 5 - 6 | 682 | 90 | |
AK161018.1 | - | 5 - 6 | 681 | 90 | ||
AY313606.1 | - | 5 - 6 | 817 | 90 | ||
BC052487.1 | - | 5 - 6 | 641 | 90 | ||
BC094340.1 | - | 5 - 6 | 895 | 30609151 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |