Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wrap73 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021499.2 | NP_067474.2 | 3 - 4 | 414 | 93 | |
GenBank mRNA | AB034911.2 | - | 3 - 4 | 394 | 93 | |
AK011773.1 | - | 3 - 4 | 378 | 93 | ||
AK051688.1 | - | 3 - 4 | 358 | 93 | ||
AK156969.1 | - | 3 - 4 | 413 | 93 | ||
AK160331.1 | - | 3 - 4 | 386 | 93 | ||
AK172690.1 | - | 3 - 4 | 413 | 93 | ||
BC016120.1 | - | 3 - 4 | 377 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |