Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Camta1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081557.3 | NP_001075026.1 | 5 - 6 | 537 | 85 | |
GenBank mRNA | BC150740.1 | - | 4 - 5 | 386 | 9087651 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |