Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Noc2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021303.2 | NP_067278.2 | 6 - 7 | 753 | 91 | |
GenBank mRNA | AK076287.1 | - | 5 - 6 | 795 | 91 | |
AK133834.1 | - | 6 - 7 | 753 | 91 | ||
AK157765.1 | - | 6 - 7 | 745 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mus musculus Probe spans exons |