Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sox5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113559.2 | NP_001107031.2 | 2 - 3 | 476 | 124 | |
NM_011444.3 | NP_035574.2 | 2 - 3 | 476 | 124 | ||
GenBank mRNA | AB006330.1 | - | 2 - 3 | 388 | 124 | |
AK029047.1 | - | 5 - 6 | 621 | 124 | ||
AK132307.1 | - | 2 - 3 | 369 | 124 | ||
BC110478.2 | - | 2 - 3 | 360 | 40047865 | 124 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |