Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plac8l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027072.1 | NP_081348.1 | 1 - 2 | 207 | 85 | |
GenBank mRNA | AK005800.1 | - | 1 - 2 | 208 | 85 | |
AK006002.1 | - | 1 - 2 | 185 | 85 | ||
BC110558.1 | - | 1 - 2 | 169 | 40049443 | 85 | |
BC110559.1 | - | 1 - 2 | 169 | 40049451 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |