Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkaa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178143.2 | NP_835279.2 | 4 - 5 | 574 | 88 | |
GenBank mRNA | AK044030.1 | - | 4 - 5 | 410 | 88 | |
AK134573.1 | - | 4 - 5 | 432 | 88 | ||
BC138565.1 | - | 4 - 5 | 583 | 8861585 | 88 | |
BC138566.1 | - | 4 - 5 | 583 | 8861586 | 88 | |
BY718361.1 | - | 4 - 5 | 429 | 88 | ||
FJ588504.1 | - | 3 - 4 | 332 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protein Kinase Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |