Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plekhn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008233.3 | NP_001008234.2 | 4 - 5 | 462 | 63 | |
GenBank mRNA | AK048084.1 | - | 2 - 3 | 2441 | 63 | |
BC046544.1 | - | 4 - 5 | 462 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |