Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dgkb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178681.4 | NP_848796.2 | 9 - 10 | 1241 | 75 | |
GenBank mRNA | AK038544.1 | - | 9 - 10 | 969 | 75 | |
AK044516.1 | - | 9 - 10 | 1051 | 75 | ||
AK047682.1 | - | 10 - 11 | 1177 | 75 | ||
AK083222.1 | - | 10 - 11 | 1252 | 75 | ||
AK122355.1 | - | 8 - 9 | 729 | 75 | ||
AK139408.1 | - | 9 - 10 | 1242 | 75 | ||
BC070461.1 | - | 9 - 10 | 1073 | 30649561 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |