Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400006571.mm.1 |
NM_001005784.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400006571.mm.1 |
NM_001005784.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400006571.mm.1 |
NM_172696.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400006571.mm.1 |
NM_172696.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400006571.mm.1 |
TC0400000982.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400006575.mm.1 |
NM_001005784.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400006575.mm.1 |
NM_001005784.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400006575.mm.1 |
NM_172696.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400006575.mm.1 |
NM_172696.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400006575.mm.1 |
TC0400000982.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400006576.mm.1 |
NM_001005784.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400006576.mm.1 |
NM_001005784.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400006576.mm.1 |
NM_172696.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400006576.mm.1 |
NM_172696.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400006576.mm.1 |
TC0400000982.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400006569.mm.1 |
NM_001005784.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400006569.mm.1 |
NM_001005784.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400006569.mm.1 |
NM_172696.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400006569.mm.1 |
NM_172696.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400006569.mm.1 |
TC0400000982.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400006570.mm.1 |
NM_001005784.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400006570.mm.1 |
NM_001005784.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400006570.mm.1 |
NM_172696.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400006570.mm.1 |
NM_172696.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400006570.mm.1 |
TC0400000982.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400006572.mm.1 |
NM_001005784.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400006572.mm.1 |
NM_001005784.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400006572.mm.1 |
NM_172696.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400006572.mm.1 |
NM_172696.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400006572.mm.1 |
TC0400000982.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400006579.mm.1 |
NM_001005784.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400006579.mm.1 |
NM_001005784.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400006579.mm.1 |
NM_172696.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400006579.mm.1 |
NM_172696.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400006579.mm.1 |
TC0400000982.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400006574.mm.1 |
NM_001005784.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400006574.mm.1 |
NM_001005784.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400006574.mm.1 |
NM_172696.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400006574.mm.1 |
NM_172696.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400006574.mm.1 |
TC0400000982.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400006580.mm.1 |
NM_001005784.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400006580.mm.1 |
NM_001005784.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400006580.mm.1 |
NM_172696.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400006580.mm.1 |
NM_172696.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400006580.mm.1 |
TC0400000982.mm.1 |
MTA TC |