Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hebp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013546.2 | NP_038574.2 | 1 - 2 | 190 | 68 | |
GenBank mRNA | AA022130.1 | - | 178 | 68 | ||
AB013095.1 | - | 1 - 2 | 129 | 68 | ||
AF117613.1 | - | 1 - 2 | 178 | 68 | ||
AK008919.1 | - | 1 - 2 | 111 | 68 | ||
BC012654.1 | - | 1 - 2 | 114 | 4216585 | 68 | |
CT010262.1 | - | 1 - 2 | 72 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |