Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Txnip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009935.2 | NP_001009935.1 | 3 - 4 | 760 | 94 | |
NM_023719.2 | NP_076208.2 | 3 - 4 | 757 | 94 | ||
GenBank mRNA | AF282826.1 | - | 3 - 4 | 478 | 94 | |
AK004653.2 | - | 3 - 4 | 757 | 94 | ||
AK082476.1 | - | 3 - 4 | 755 | 94 | ||
AK088870.1 | - | 3 - 4 | 755 | 94 | ||
AK089304.1 | - | 3 - 4 | 760 | 94 | ||
AK089351.1 | - | 3 - 4 | 758 | 94 | ||
AK089403.1 | - | 3 - 4 | 760 | 94 | ||
AK155517.1 | - | 3 - 4 | 758 | 94 | ||
AK171205.1 | - | 3 - 4 | 758 | 94 | ||
AK171312.1 | - | 3 - 4 | 758 | 94 | ||
AK171425.1 | - | 3 - 4 | 758 | 94 | ||
AK171456.1 | - | 3 - 4 | 755 | 94 | ||
AK171464.1 | - | 3 - 4 | 758 | 94 | ||
AK171527.1 | - | 3 - 4 | 758 | 94 | ||
AK192038.1 | - | 3 - 4 | 355 | 94 | ||
AK214578.1 | - | 3 - 4 | 332 | 94 | ||
BC011212.1 | - | 3 - 4 | 4193517 | 94 | ||
BC031850.1 | - | 3 - 4 | 3591421 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |