Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nudt17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001162925.1 | NP_001156397.1 | 4 - 5 | 591 | 66 | |
NM_030094.1 | NP_084370.1 | 4 - 5 | 540 | 66 | ||
GenBank mRNA | AK019109.1 | - | 4 - 5 | 545 | 66 | |
AK134929.1 | - | 4 - 5 | 528 | 66 | ||
AK141256.1 | - | 4 - 5 | 528 | 66 | ||
BC115559.2 | - | 4 - 5 | 576 | 40101853 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |