Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lix1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163170.1 | NP_001156642.1 | 2 - 3 | 503 | 80 | |
GenBank mRNA | AK051276.1 | - | 2 - 3 | 488 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mus musculus Probe spans exons |