Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081139.1 | NP_001074608.1 | 3 - 4 | 541 | 87 | |
GenBank mRNA | AK028691.1 | - | 3 - 4 | 331 | 87 | |
AK039772.1 | - | 3 - 4 | 296 | 87 | ||
AK046624.1 | - | 3 - 4 | 350 | 87 | ||
BC138276.1 | - | 3 - 4 | 343 | 8861295 | 87 | |
BC145228.1 | - | 3 - 4 | 347 | 9053780 | 87 | |
BC145750.1 | - | 3 - 4 | 343 | 40131053 | 87 | |
BC157973.1 | - | 3 - 4 | 343 | 9007295 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Apoptosis Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |