Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prdm16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001177995.1 | NP_001171466.1 | 5 - 6 | 790 | 68 | |
NM_001291026.1 | NP_001277955.1 | 5 - 6 | 784 | 68 | ||
NM_001291029.1 | NP_001277958.1 | 5 - 6 | 790 | 68 | ||
NM_027504.3 | NP_081780.3 | 5 - 6 | 787 | 68 | ||
GenBank mRNA | AB078338.1 | - | 5 - 6 | 679 | 68 | |
AK083829.1 | - | 5 - 6 | 674 | 68 | ||
BC053082.1 | - | 5 - 6 | 725 | 68 | ||
BC059838.1 | - | 5 - 6 | 768 | 6409778 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |