Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001146196.1 | NP_001139668.1 | 4 - 5 | 782 | 77 | |
NM_009132.2 | NP_033158.2 | 4 - 5 | 782 | 77 | ||
GenBank mRNA | AK133484.1 | - | 4 - 5 | 738 | 77 | |
AK136080.1 | - | 2 - 3 | 324 | 77 | ||
AK167739.1 | - | 4 - 5 | 737 | 77 | ||
AK173295.1 | - | 4 - 5 | 744 | 77 | ||
BC063328.1 | - | 4 - 5 | 737 | 30314464 | 77 | |
U04354.1 | - | 4 - 5 | 737 | 77 | ||
Y13971.1 | - | 4 - 5 | 737 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |