Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cep76 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081073.1 | NP_001074542.1 | 4 - 5 | 683 | 159 | |
GenBank mRNA | AK035879.1 | - | 3 - 4 | 463 | 159 | |
BC119216.1 | - | 4 - 5 | 564 | 8733965 | 159 | |
BC119218.1 | - | 4 - 5 | 564 | 8733967 | 159 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |