Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: St8sia1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011374.2 | NP_035504.2 | 1 - 2 | 921 | 77 | |
GenBank mRNA | AK046188.1 | - | 1 - 2 | 861 | 77 | |
AK052444.1 | - | 1 - 2 | 572 | 77 | ||
BC024821.1 | - | 1 - 2 | 921 | 5357151 | 77 | |
L38677.1 | - | 1 - 2 | 545 | 77 | ||
X84235.1 | - | 1 - 2 | 268 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |