Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: St8sia1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011374.2 | NP_035504.2 | 2 - 3 | 1063 | 73 | |
GenBank mRNA | AK042207.1 | - | 2 - 3 | 919 | 73 | |
AK046188.1 | - | 2 - 3 | 1003 | 73 | ||
AK052444.1 | - | 2 - 3 | 714 | 73 | ||
BC024821.1 | - | 2 - 3 | 1063 | 5357151 | 73 | |
EF101558.1 | - | 2 - 3 | 175 | 73 | ||
L38677.1 | - | 2 - 3 | 687 | 73 | ||
X84235.1 | - | 2 - 3 | 410 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |