Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1200002873.mm.1 |
NM_001015099.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200002873.mm.1 |
NM_001015099.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200002873.mm.1 |
NM_001167963.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200002873.mm.1 |
NM_001167963.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200002873.mm.1 |
NM_001167964.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200002873.mm.1 |
NM_001167964.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200002873.mm.1 |
TC1200000415.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200002874.mm.1 |
NM_001015099.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200002874.mm.1 |
NM_001015099.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200002874.mm.1 |
NM_001167963.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200002874.mm.1 |
NM_001167963.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200002874.mm.1 |
NM_001167964.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200002874.mm.1 |
NM_001167964.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200002874.mm.1 |
TC1200000415.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200002877.mm.1 |
NM_001015099.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200002877.mm.1 |
NM_001015099.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200002877.mm.1 |
NM_001167963.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200002877.mm.1 |
NM_001167963.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200002877.mm.1 |
NM_001167964.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200002877.mm.1 |
NM_001167964.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200002877.mm.1 |
TC1200000415.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200002876.mm.1 |
NM_001015099.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200002876.mm.1 |
NM_001015099.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200002876.mm.1 |
NM_001167963.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200002876.mm.1 |
NM_001167963.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200002876.mm.1 |
NM_001167964.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200002876.mm.1 |
NM_001167964.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200002876.mm.1 |
TC1200000415.mm.1 |
MTA TC |