Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Meis3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK083907.1 | - | 1 - 2 | 180 | 111 | |
AK142345.1 | - | 1 - 2 | 164 | 111 | ||
AK155364.1 | - | 1 - 2 | 170 | 111 | ||
BC003762.1 | - | 1 - 2 | 161 | 3593200 | 111 | |
BC117532.1 | - | 1 - 2 | 82 | 40106322 | 111 | |
BC118513.1 | - | 1 - 2 | 82 | 40106324 | 111 | |
BY273565.1 | - | 1 - 2 | 242 | 111 | ||
BY291772.1 | - | 1 - 2 | 293 | 111 | ||
U57344.1 | - | 1 - 2 | 137 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Mus musculus 1700 Probe spans exons |