Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700021335.mm.1 |
TC0700002424.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700021335.mm.1 |
NM_001136471.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700021335.mm.1 |
NM_001136471.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700021335.mm.1 |
NM_172297.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700021335.mm.1 |
NM_172297.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700021334.mm.1 |
TC0700002424.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700021334.mm.1 |
NM_001136471.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700021334.mm.1 |
NM_001136471.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700021334.mm.1 |
NM_172297.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700021334.mm.1 |
NM_172297.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700021330.mm.1 |
TC0700002424.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700021330.mm.1 |
NM_001136471.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700021330.mm.1 |
NM_001136471.1 |
Refseq |