Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc8a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_148946.2 | NP_683748.1 | 4 - 5 | 1899 | 109 | |
GenBank mRNA | AF503502.1 | - | 3 - 4 | 1764 | 109 | |
AK035163.1 | - | 4 - 5 | 2041 | 109 | ||
BC058704.1 | - | 4 - 5 | 1899 | 6838206 | 109 | |
BC080277.1 | - | 4 - 5 | 1851 | 30354551 | 109 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |