Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aspscr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164224.1 | NP_001157696.1 | 11 - 12 | 1203 | 62 | |
NM_026877.2 | NP_081153.1 | 13 - 14 | 1487 | 62 | ||
NM_198223.2 | NP_937866.1 | 12 - 13 | 1266 | 62 | ||
GenBank mRNA | AK004509.1 | - | 7 - 8 | 499 | 62 | |
AK088197.1 | - | 13 - 14 | 1325 | 62 | ||
AK146061.1 | - | 11 - 12 | 1204 | 62 | ||
AY349132.1 | - | 11 - 12 | 1256 | 62 | ||
AY349133.1 | - | 13 - 14 | 1404 | 62 | ||
BC019177.1 | - | 13 - 14 | 1430 | 5008127 | 62 | |
BC022115.1 | - | 13 - 14 | 1434 | 5101238 | 62 | |
BC031153.1 | - | 13 - 14 | 1419 | 4976569 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |