Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aspscr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026877.2 | NP_081153.1 | 2 - 3 | 259 | 69 | |
GenBank mRNA | AY349133.1 | - | 2 - 3 | 176 | 69 | |
BC019177.1 | - | 2 - 3 | 202 | 5008127 | 69 | |
BC022115.1 | - | 2 - 3 | 206 | 5101238 | 69 | |
BC031153.1 | - | 2 - 3 | 191 | 4976569 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |