Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nop56 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024193.2 | NP_077155.2 | 3 - 4 | 269 | 60 | |
GenBank mRNA | AK009799.1 | - | 3 - 4 | 255 | 60 | |
AK011481.1 | - | 3 - 4 | 236 | 60 | ||
AK011744.1 | - | 3 - 4 | 234 | 60 | ||
AK013528.1 | - | 3 - 4 | 236 | 60 | ||
AK030730.1 | - | 3 - 4 | 238 | 60 | ||
AK077795.1 | - | 3 - 4 | 236 | 60 | ||
AK139924.1 | - | 3 - 4 | 236 | 60 | ||
AK146243.1 | - | 3 - 4 | 236 | 60 | ||
AK150258.1 | - | 3 - 4 | 236 | 60 | ||
AK154024.1 | - | 3 - 4 | 237 | 60 | ||
BC021355.1 | - | 3 - 4 | 231 | 5006404 | 60 | |
CJ162437.1 | - | 3 - 4 | 272 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |