Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tspan32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001128080.2 | NP_001121552.1 | 6 - 7 | 611 | 76 | |
NM_020286.3 | NP_064682.1 | 5 - 6 | 726 | 76 | ||
GenBank mRNA | AF291425.1 | - | 5 - 6 | 694 | 76 | |
AJ279791.1 | - | 5 - 6 | 686 | 76 | ||
AK154694.1 | - | 5 - 6 | 747 | 76 | ||
AY039000.1 | - | 6 - 7 | 619 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons |