Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175173.3 | NP_780382.2 | 21 - 22 | 2155 | 67 | |
GenBank mRNA | AK040117.1 | - | 6 - 7 | 595 | 67 | |
AK077050.1 | - | 21 - 22 | 2156 | 67 | ||
AV277048.2 | - | 1 - 2 | 73 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus Probe spans exons |