Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zmiz1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_183208.3 | NP_899031.2 | 20 - 21 | 3052 | 119 | |
GenBank mRNA | BC057691.1 | - | 10 - 11 | 1289 | 119 | |
BC058646.1 | - | 20 - 21 | 2952 | 6405777 | 119 | |
BC065120.1 | - | 20 - 21 | 2945 | 6856060 | 119 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |