Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clic5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172621.2 | NP_766209.1 | 1 - 2 | 392 | 61 | |
GenBank mRNA | AK046522.1 | - | 1 - 2 | 381 | 61 | |
AK156849.1 | - | 1 - 2 | 233 | 61 | ||
BC064037.1 | - | 1 - 2 | 377 | 1227040 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel Mammalian Gene Collection Mus musculus Probe spans exons |