Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crisp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204071.1 | NP_001191000.1 | 1 - 2 | 120 | 59 | |
NM_009420.2 | NP_033446.1 | 1 - 2 | 120 | 59 | ||
GenBank mRNA | AK018856.1 | - | 1 - 2 | 51 | 59 | |
BC049615.1 | - | 1 - 2 | 72 | 6742033 | 59 | |
CJ046590.1 | - | 1 - 2 | 61 | 59 | ||
M25533.1 | - | 1 - 2 | 72 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Mus musculus Probe spans exons |