Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kat6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205241.1 | NP_001192170.1 | 4 - 5 | 1150 | 93 | |
NM_017479.3 | NP_059507.2 | 4 - 5 | 1274 | 93 | ||
GenBank mRNA | AF222800.1 | - | 4 - 5 | 1190 | 93 | |
AK045188.1 | - | 4 - 5 | 1286 | 93 | ||
AK135465.1 | - | 4 - 5 | 1326 | 93 | ||
AK147509.1 | - | 4 - 5 | 1151 | 93 | ||
BC095974.1 | - | 3 - 4 | 927 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |